The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009712.3 | NP_033842.3 | 1 - 2 | 413 | 109 |
GenBank mRNA | AK083309.1 | - | 1 - 2 | 397 | 109 |
AK140163.1 | - | 1 - 2 | 463 | 109 | |
AK154098.1 | - | 1 - 2 | 333 | 109 | |
AK158312.1 | - | 1 - 2 | 414 | 109 | |
BC150661.1 | - | 1 - 2 | 370 | 109 |