The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023821.3 | NP_076310.2 | 12 - 13 | 11000 | 99 |
GenBank mRNA | AJ250188.1 | - | 3 - 4 | 373 | 99 |
AJ511265.1 | - | 11 - 12 | 10677 | 99 | |
AJ575748.1 | - | 12 - 13 | 11189 | 99 | |
AK010194.1 | - | 4 - 5 | 486 | 99 |