The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048267.1 | NP_001041732.1 | 23 - 24 | 2843 | 122 |
NM_178716.3 | NP_848831.2 | 23 - 24 | 2849 | 122 | |
GenBank mRNA | AK029953.1 | - | 23 - 24 | 2850 | 122 |
AK030580.1 | - | 23 - 24 | 2809 | 122 | |
AK045132.1 | - | 23 - 24 | 2843 | 122 | |
AK163279.1 | - | 6 - 7 | 512 | 122 | |
BC031571.1 | - | 15 - 16 | 1892 | 122 | |
BC055372.1 | - | 23 - 24 | 2750 | 122 |