The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028120.2 | NP_082396.1 | 14 - 15 | 1644 | 77 |
GenBank mRNA | AK012067.1 | - | 14 - 15 | 1645 | 77 |
AK029107.1 | - | 14 - 15 | 1675 | 77 | |
BC003981.1 | - | 14 - 15 | 1699 | 77 |