Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samd7
Entrez Gene ID
75953
Gene Name
sterile alpha motif domain containing 7
Gene Aliases
4930597A01Rik
Chromosome Location
Chr.3: 30746293 - 30767174 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029489.3 NP_083765.2 3 - 4 342 65
GenBank mRNA DV659349.1 - 3 - 4 295 65

View Full Product Details