Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sptb
Entrez Gene ID
20741
Gene Name
spectrin beta, erythrocytic
Gene Aliases
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219
Chromosome Location
Chr.12: 76580488 - 76710547 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013675.3 NP_038703.3 34 - 35 7449 107
GenBank mRNA AK147299.1 - 34 - 35 7450 107
AK147684.1 - 34 - 35 7042 107
AK147704.1 - 34 - 35 7042 107
AK220551.1 - 15 - 16 2549 107
BC117711.2 - 17 - 18 2703 107
BC129806.1 - 33 - 34 6782 107
BC150783.1 - 33 - 34 6815 107

View Full Product Details