Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sptb
Entrez Gene ID
20741
Gene Name
spectrin beta, erythrocytic
Gene Aliases
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219
Chromosome Location
Chr.12: 76580488 - 76710547 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013675.3 NP_038703.3 8 - 9 1542 124
GenBank mRNA AK147299.1 - 8 - 9 1543 124
AK147684.1 - 8 - 9 1135 124
AK147704.1 - 8 - 9 1135 124
BC129806.1 - 7 - 8 875 124
BC150783.1 - 7 - 8 908 124
S66283.1 - 8 - 9 1018 124

View Full Product Details