Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Dcun1d1
- Entrez Gene ID
-
114893
- Gene Name
- DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae)
- Gene Aliases
-
Rp42, SCCRO, Tes3, pTes3
- Chromosome Location
-
Chr.3: 35892103 - 35937605 on Build GRCm38
- Gene Symbol
- Tes3-ps
- Entrez Gene ID
-
54339
- Gene Name
- testis derived transcript 3, pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 49493652 - 49496389 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK029314.1 |
- |
1 - 1 |
556 |
107 |
View Full Product Details