The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163516.2 | NP_001156988.1 | 12 - 13 | 1820 | 61 |
NM_001163517.2 | NP_001156989.1 | 7 - 8 | 942 | 61 | |
NM_001289505.1 | NP_001276434.1 | 14 - 15 | 1801 | 61 | |
NM_021483.4 | NP_067458.2 | 14 - 15 | 1930 | 61 | |
GenBank mRNA | AB032591.1 | - | 14 - 15 | 1925 | 61 |
AK006035.1 | - | 7 - 8 | 943 | 61 | |
AK039764.1 | - | 14 - 15 | 1800 | 61 | |
AK083141.1 | - | 12 - 13 | 1712 | 61 | |
HQ646186.1 | - | 15 - 16 | 2149 | 61 | |
HQ646187.1 | - | 14 - 15 | 2083 | 61 | |
HQ646188.1 | - | 13 - 14 | 1978 | 61 | |
HQ646189.1 | - | 13 - 14 | 2011 | 61 | |
HQ646190.1 | - | 12 - 13 | 1906 | 61 | |
HQ646191.1 | - | 14 - 15 | 2739 | 61 | |
HQ646192.1 | - | 15 - 16 | 2153 | 61 | |
HQ646193.1 | - | 15 - 16 | 2059 | 61 | |
HQ646194.1 | - | 14 - 15 | 1954 | 61 | |
HQ646195.1 | - | 15 - 16 | 1927 | 61 | |
HQ646196.1 | - | 14 - 15 | 1822 | 61 | |
HQ646197.1 | - | 7 - 8 | 942 | 61 |