Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rufy1
Entrez Gene ID
216724
Gene Name
RUN and FYVE domain containing 1
Gene Aliases
3000002E04Rik, Rabip4, ZFYVE12
Chromosome Location
Chr.11: 50389286 - 50431111 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172557.2 NP_766145.1 15 - 16 1874 81
GenBank mRNA AJ250024.1 - 14 - 15 1550 81
AK046633.1 - 15 - 16 1875 81

View Full Product Details