Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rufy1
Entrez Gene ID
216724
Gene Name
RUN and FYVE domain containing 1
Gene Aliases
3000002E04Rik, Rabip4, ZFYVE12
Chromosome Location
Chr.11: 50389286 - 50431111 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172557.2 NP_766145.1 17 - 18 2000 76
GenBank mRNA AJ250024.1 - 16 - 17 1676 76
AK046633.1 - 17 - 18 2001 76

View Full Product Details