Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sell
Entrez Gene ID
20343
Gene Name
selectin, lymphocyte
Gene Aliases
AI528707, CD62L, L-selectin, LECAM-1, Lnhr, Ly-22, Ly-m22, Lyam-1, Lyam1
Chromosome Location
Chr.1: 164061813 - 164080785 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011346.2 NP_035476.1 3 - 4 585 72
GenBank mRNA AB353767.1 - 2 - 3 472 72
AB353768.1 - 2 - 3 472 72
AK137509.1 - 3 - 4 497 72
AK157273.1 - 3 - 4 574 72
AK161754.1 - 3 - 4 72
AK163066.1 - 1 - 2 42 72
AK172116.1 - 3 - 4 575 72
BC052681.1 - 3 - 4 569 72
M25324.1 - 3 - 4 577 72
M36005.1 - 3 - 4 523 72
M36058.1 - 3 - 4 523 72
X14772.1 - 3 - 4 526 72

View Full Product Details