The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020614.2 | NP_065639.2 | 6 - 7 | 597 | 95 |
NR_028341.1 | 5 - 6 | 499 | 95 | ||
GenBank mRNA | AK015111.1 | - | 6 - 7 | 564 | 95 |
AK131963.1 | - | 6 - 7 | 571 | 95 | |
AK165580.1 | - | 6 - 7 | 577 | 95 | |
AK170313.1 | - | 6 - 7 | 563 | 95 | |
BC057167.1 | - | 6 - 7 | 584 | 95 | |
BC069907.1 | - | 5 - 6 | 454 | 95 | |
BC086643.1 | - | 6 - 7 | 560 | 95 | |
BC094035.1 | - | 5 - 6 | 449 | 95 | |
BC125027.1 | - | 6 - 7 | 546 | 95 | |
BC125028.1 | - | 6 - 7 | 546 | 95 | |
CN721617.1 | - | 1 - 2 | 158 | 95 |