Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp1r12b
Entrez Gene ID
329251
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 12B
Gene Aliases
1810037O03Rik, 9530009M10Rik, AI132431, Mypt2
Chromosome Location
Chr.1: 134754665 - 134955965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081307.1 NP_001074776.1 15 - 16 2280 61
GenBank mRNA AK039445.1 - 3 - 4 341 61
AK141375.1 - 17 - 18 2495 61
BC145746.1 - 15 - 16 2191 61

View Full Product Details