The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128091.1 | NP_001121563.1 | 4 - 5 | 841 | 92 |
GenBank mRNA | BC147724.1 | - | 4 - 5 | 841 | 92 |
BC147728.1 | - | 4 - 5 | 841 | 92 |