Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Itk
Entrez Gene ID
16428
Gene Name
IL2 inducible T cell kinase
Gene Aliases
Emt, Tcsk, Tsk
Chromosome Location
Chr.11: 46325148 - 46389515 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281965.1 NP_001268894.1 16 - 17 1905 66
NM_001281966.1 NP_001268895.1 16 - 17 1896 66
NM_001281968.1 NP_001268897.1 16 - 17 1884 66
NM_010583.3 NP_034713.2 16 - 17 1887 66
GenBank mRNA AK037554.1 - - 967 66
AK083380.1 - 16 - 17 1859 66
AK153613.1 - 16 - 17 1843 66
BC128374.1 - 16 - 17 1813 66
BC128375.1 - 16 - 17 1810 66
D14042.1 - 16 - 17 1827 66
L00619.1 - 16 - 17 1892 66
L05631.1 - 16 - 17 1853 66
L10628.1 - 16 - 17 1855 66

View Full Product Details