Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Espnl
Entrez Gene ID
227357
Gene Name
espin-like
Gene Aliases
Gm556
Chromosome Location
Chr.1: 91321164 - 91348303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033292.3 NP_001028464.1 3 - 4 734 60
GenBank mRNA AK134448.1 - 3 - 4 736 60

View Full Product Details