Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam20b
Entrez Gene ID
215015
Gene Name
family with sequence similarity 20, member B
Gene Aliases
C530043G21Rik, mKIAA0475
Chromosome Location
Chr.1: 156678545 - 156719027 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145413.4 NP_663388.1 2 - 3 543 107
GenBank mRNA AK049705.1 - 1 - 2 345 107
AK132149.1 - 2 - 3 544 107
BC019381.1 - 2 - 3 562 107
BC023737.1 - 3 - 4 759 107
BC024412.1 - 1 - 2 493 107
BC031473.1 - 2 - 3 655 107

View Full Product Details