The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145413.4 | NP_663388.1 | 4 - 5 | 747 | 64 |
GenBank mRNA | AK049705.1 | - | 3 - 4 | 549 | 64 |
AK129154.1 | - | 2 - 3 | 156 | 64 | |
AK132149.1 | - | 4 - 5 | 748 | 64 | |
BC019381.1 | - | 4 - 5 | 766 | 64 | |
BC023737.1 | - | 5 - 6 | 963 | 64 | |
BC024412.1 | - | 3 - 4 | 697 | 64 | |
BC031473.1 | - | 4 - 5 | 859 | 64 |