Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajc19
Entrez Gene ID
67713
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 19
Gene Aliases
1810055D05Rik, AA959924, Tim14
Chromosome Location
Chr.3: 34057280 - 34081354 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026211.2 NP_001021382.1 1 - 2 169 78
NM_001286972.1 NP_001273901.1 1 - 2 169 78
NM_026332.4 NP_080608.3 1 - 2 169 78
GenBank mRNA AK008267.1 - 1 - 2 113 78
AK010204.1 - 1 - 2 119 78
AK012051.1 - 1 - 2 121 78
AK076602.1 - - 128 78
BC020327.1 - 1 - 2 102 78
BC024335.1 - 1 - 2 77 78
BY149679.1 - 1 - 2 142 78
CA469052.1 - 1 - 2 206 78
EL962106.1 - 1 - 2 115 78

View Full Product Details