Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Efhc2
Entrez Gene ID
74405
Gene Name
EF-hand domain (C-terminal) containing 2
Gene Aliases
4933407D04Rik, mRib72-2
Chromosome Location
Chr.X: 17132049 - 17320461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028916.4 NP_083192.2 13 - 14 2119 128
GenBank mRNA AK016714.1 - 13 - 14 2078 128
BC120659.1 - 13 - 14 2054 128
BC125625.1 - 13 - 14 2054 128
BE956904.1 - 2 - 3 128

View Full Product Details