Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp9x
Entrez Gene ID
22284
Gene Name
ubiquitin specific peptidase 9, X chromosome
Gene Aliases
5730589N07Rik, AA407302, AA407699, AL022658, AL022749, Dffrx, FAF-X, Fafl
Chromosome Location
Chr.X: 13070997 - 13173328 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009481.2 NP_033507.2 30 - 31 5003 117
GenBank mRNA DQ086491.1 - 30 - 31 4964 117
U67874.1 - 30 - 31 4964 117

View Full Product Details