The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009481.2 | NP_033507.2 | 4 - 5 | 713 | 125 |
GenBank mRNA | AK028443.1 | - | 4 - 5 | 698 | 125 |
AK049283.1 | - | 5 - 6 | 826 | 125 | |
DQ086491.1 | - | 4 - 5 | 674 | 125 | |
U67874.1 | - | 4 - 5 | 674 | 125 |