The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163328.1 | NP_001156800.1 | 2 - 3 | 567 | 81 |
NM_178631.4 | NP_848746.1 | 3 - 4 | 660 | 81 | |
GenBank mRNA | AK032086.1 | - | 2 - 3 | 500 | 81 |
AK079659.1 | - | 2 - 3 | 1389 | 81 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK003003.1 | - | Chr. - - [-] on Build | 80 |