The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163579.1 | NP_001157051.1 | 9 - 10 | 1544 | 105 |
NM_001163580.1 | NP_001157052.1 | 9 - 10 | 1496 | 105 | |
NM_028915.3 | NP_083191.2 | 9 - 10 | 1544 | 105 | |
GenBank mRNA | AK004646.1 | - | 9 - 10 | 1545 | 105 |
AK016559.1 | - | 9 - 10 | 1483 | 105 | |
AK029563.1 | - | 9 - 10 | 1461 | 105 | |
AK030800.1 | - | 1 - 2 | 135 | 105 | |
AK173256.1 | - | 9 - 10 | 1472 | 105 | |
BC061202.1 | - | 9 - 10 | 1452 | 105 | |
BC099680.1 | - | 9 - 10 | 1459 | 105 | |
BC138734.1 | - | 9 - 10 | 1518 | 105 | |
BC138735.1 | - | 9 - 10 | 1518 | 105 |