Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gcc2
Entrez Gene ID
70297
Gene Name
GRIP and coiled-coil domain containing 2
Gene Aliases
0610043A03Rik, 2210420P05Rik, 2600014C01Rik, AW112121, mKIAA0336
Chromosome Location
Chr.10: 58254296 - 58305599 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027375.2 NP_081651.2 6 - 7 2794 101
GenBank mRNA AB093232.1 - 6 - 7 2796 102
BC023811.1 - 1 - 2 181 101
BC139007.1 - 6 - 7 2790 101
BC145495.1 - 6 - 7 2863 101

View Full Product Details