The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008551.1 | NP_032577.1 | 4 - 5 | 539 | 75 |
GenBank mRNA | BC063064.1 | - | 4 - 5 | 539 | 75 |
X76850.1 | - | 4 - 5 | 518 | 75 |