Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdlim4
Entrez Gene ID
30794
Gene Name
PDZ and LIM domain 4
Gene Aliases
Ril
Chromosome Location
Chr.11: 54054928 - 54069032 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019417.3 NP_062290.2 2 - 3 327 84
GenBank mRNA AB221672.1 - 2 - 3 242 84
AK035513.1 - 2 - 3 302 84
BC030068.1 - 2 - 3 256 84
Y08361.1 - 2 - 3 278 84

View Full Product Details