Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasgef1c
Entrez Gene ID
74563
Gene Name
RasGEF domain family, member 1C
Gene Aliases
9130006A14Rik
Chromosome Location
Chr.11: 49901816 - 49980223 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029004.1 NP_083280.1 13 - 14 1516 68
GenBank mRNA AK018593.1 - 13 - 14 1505 68
AK039330.1 - - 1880 68
AK047088.1 - 12 - 13 1395 68

View Full Product Details