The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029004.1 | NP_083280.1 | 3 - 4 | 443 | 82 |
GenBank mRNA | AK018593.1 | - | 3 - 4 | 432 | 82 |
AK039330.1 | - | - | 807 | 82 |