The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001131054.1 | NP_001124526.1 | 4 - 5 | 414 | 78 |
NM_013917.2 | NP_038945.2 | 3 - 4 | 666 | 78 | |
GenBank mRNA | AK002473.1 | - | 3 - 4 | 380 | 78 |
AK008704.1 | - | 3 - 4 | 381 | 78 | |
AK088387.1 | - | 3 - 4 | 380 | 78 | |
CN837307.1 | - | 4 - 5 | 419 | 78 | |
DV657646.1 | - | 3 - 4 | 384 | 78 |