Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elk4
Entrez Gene ID
13714
Gene Name
ELK4, member of ETS oncogene family
Gene Aliases
2310011G17Rik, A130026I01Rik, BB162516, SAP-1, Sap1
Chromosome Location
Chr.1: 132007234 - 132032612 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC004798.1 - 3 - 4 1073 70
BC041679.1 - 3 - 4 1061 70

View Full Product Details