Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnmb2
Entrez Gene ID
72413
Gene Name
potassium large conductance calcium-activated channel, subfamily M, beta member 2
Gene Aliases
2700049B16Rik, 3110031N04Rik
Chromosome Location
Chr.3: 31902132 - 32200180 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028231.2 NP_082507.1 1 - 2 225 78
GenBank mRNA BC046227.1 - 1 - 2 225 78
BC058957.1 - 1 - 2 225 78

View Full Product Details