Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrriq4
Entrez Gene ID
68307
Gene Name
leucine-rich repeats and IQ motif containing 4
Gene Aliases
4930558O21Rik
Chromosome Location
Chr.3: 30644506 - 30672431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290510.1 NP_001277439.1 3 - 4 1592 150
NM_026668.2 NP_080944.2 5 - 6 1755 150
GenBank mRNA AK015024.1 - 3 - 4 1543 150
AK019775.1 - 5 - 6 1756 150
BC145806.1 - 5 - 6 1456 150

View Full Product Details