Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tssc1
Entrez Gene ID
380752
Gene Name
tumor suppressing subtransferable candidate 1
Gene Aliases
D12Ertd604e
Chromosome Location
Chr.12: 28751804 - 28867494 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201357.2 NP_958745.2 8 - 9 1116 116
GenBank mRNA AK150308.1 - 8 - 9 1117 116
AK151830.1 - 8 - 9 1117 116
BC031458.1 - 8 - 9 1079 116

View Full Product Details