The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205067.1 | NP_001191996.1 | 6 - 7 | 1046 | 85 |
NM_024205.2 | NP_077167.1 | 6 - 7 | 1064 | 85 | |
GenBank mRNA | AK004555.1 | - | 6 - 7 | 1001 | 85 |
AK013502.1 | - | 1 - 2 | 93 | 85 | |
BC002238.1 | - | 6 - 7 | 780 | 85 |