Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpa33
Entrez Gene ID
59290
Gene Name
glycoprotein A33 (transmembrane)
Gene Aliases
2010310L10Rik, 2210401D16Rik, BB116197
Chromosome Location
Chr.1: 166130460 - 166166510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021610.1 NP_067623.1 2 - 3 298 139
GenBank mRNA AF247659.1 - 2 - 3 298 139
AK008784.1 - 2 - 3 330 139
BC008528.1 - 2 - 3 291 139

View Full Product Details