Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tdrd5
Entrez Gene ID
214575
Gene Name
tudor domain containing 5
Gene Aliases
Gm103
Chromosome Location
Chr.1: 156255296 - 156305295 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134741.1 NP_001128213.1 3 - 4 670 87
GenBank mRNA AK132674.1 - 3 - 4 656 87

View Full Product Details