The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005508.2 | NP_001005508.2 | 1 - 2 | 404 | 89 |
GenBank mRNA | AK031319.1 | - | 1 - 2 | 175 | 89 |
AK147757.1 | - | 1 - 2 | 411 | 89 | |
AK156351.1 | - | 1 - 2 | 393 | 89 | |
BC082573.1 | - | 1 - 2 | 352 | 89 | |
BY740863.1 | - | 1 - 2 | 411 | 89 |