Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slco6c1
Entrez Gene ID
74441
Gene Name
solute carrier organic anion transporter family, member 6c1
Gene Aliases
4933404A18Rik, C79179
Chromosome Location
Chr.1: 97059447 - 97128340 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028942.4 NP_083218.1 3 - 4 907 79
GenBank mRNA AK016647.1 - 3 - 4 855 79
AK029505.1 - 3 - 4 908 79
AK132668.1 - 3 - 4 858 79
AK161249.1 - 3 - 4 858 79
BC139066.1 - 3 - 4 810 79
BC139067.1 - 3 - 4 810 79

View Full Product Details