Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc26a3
Entrez Gene ID
13487
Gene Name
solute carrier family 26, member 3
Gene Aliases
9030623B18Rik, 9130013M11Rik, AV376035, Dra
Chromosome Location
Chr.12: 31390876 - 31473922 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021353.3 NP_067328.1 19 - 20 2392 97
GenBank mRNA AF136751.1 - 19 - 20 2310 97
BC139271.1 - 18 - 19 2227 97
BC139273.1 - 18 - 19 2227 97

View Full Product Details