Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Sox2ot
- Entrez Gene ID
-
320478
- Gene Name
- SOX2 overlapping transcript (non-protein coding)
- Gene Aliases
-
B230215L15Rik
- Chromosome Location
-
Chr.3: 34560381 - 34677993 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK045614.1 |
- |
1 - 2 |
97 |
80 |
View Full Product Details