The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145128.3 | NP_660110.2 | 7 - 8 | 1282 | 106 |
GenBank mRNA | AF474154.1 | - | 5 - 6 | 662 | 106 |
AF474155.1 | - | 5 - 6 | 725 | 106 | |
AK030402.1 | - | 7 - 8 | 1250 | 106 | |
AK036567.1 | - | 7 - 8 | 1237 | 106 | |
AK049587.1 | - | 6 - 7 | 1205 | 106 | |
BC125533.1 | - | 5 - 6 | 690 | 106 | |
BC125535.1 | - | 5 - 6 | 690 | 106 |