Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slamf9
- Entrez Gene ID
-
98365
- Gene Name
- SLAM family member 9
- Gene Aliases
-
2310026I04Rik, AI462096, CD2F-10, CD84-H1, Cd2f10, SF2001
- Chromosome Location
-
Chr.1: 172458348 - 172478421 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK076693.1 |
- |
- |
204 |
96 |
View Full Product Details