Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btnl9
Entrez Gene ID
237754
Gene Name
butyrophilin-like 9
Gene Aliases
B430208I01, Btn3, D330012D11Rik
Chromosome Location
Chr.11: 49168325 - 49187089 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY177687.1 - 4 - 5 827 94

View Full Product Details