Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim9
Entrez Gene ID
94090
Gene Name
tripartite motif-containing 9
Gene Aliases
-
Chromosome Location
Chr.12: 70244533 - 70347761 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110203.1 NP_001103673.1 7 - 8 2051 77
GenBank mRNA AK035807.1 - 7 - 8 1769 77

View Full Product Details