Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankmy1
Entrez Gene ID
241158
Gene Name
ankyrin repeat and MYND domain containing 1
Gene Aliases
4930483I10Rik, ZMYND13
Chromosome Location
Chr.1: 92859844 - 92902986 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172850.3 NP_766438.2 13 - 14 2534 60
GenBank mRNA AK029700.1 - 13 - 14 2536 60
BC120887.1 - 12 - 13 2370 60
BC120888.1 - 12 - 13 2370 60

View Full Product Details