The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010475.1 | NP_034605.1 | 5 - 6 | 742 | 85 |
GenBank mRNA | AK019406.1 | - | 2 - 3 | 260 | 85 |
AK143346.1 | - | 5 - 6 | 691 | 85 | |
BC125657.1 | - | 5 - 6 | 741 | 85 | |
BC125659.1 | - | 5 - 6 | 741 | 85 | |
X89627.1 | - | 5 - 6 | 742 | 85 |