Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcap29
Entrez Gene ID
12033
Gene Name
B cell receptor associated protein 29
Gene Aliases
AW208404, Bap29
Chromosome Location
Chr.12: 31595354 - 31634658 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164090.1 NP_001157562.1 4 - 5 429 111
NM_007530.3 NP_031556.1 3 - 4 592 111
GenBank mRNA AK090068.1 - 4 - 5 406 111
AK144754.1 - 4 - 5 403 111
BC003303.1 - 3 - 4 496 111
BC021661.1 - 3 - 4 603 111

View Full Product Details