Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkar2b
Entrez Gene ID
19088
Gene Name
protein kinase, cAMP dependent regulatory, type II beta
Gene Aliases
AI451071, AW061005, PKARIIbeta, Pkarb2, RII(beta)
Chromosome Location
Chr.12: 31958450 - 32061343 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011158.3 NP_035288.2 8 - 9 1107 138
GenBank mRNA AK041013.1 - 8 - 9 1081 138
AK081805.1 - 8 - 9 884 138
AK135621.1 - 8 - 9 1107 138
AK138963.1 - 8 - 9 1073 138
BC033267.1 - 4 - 5 395 138

View Full Product Details